Skip to Content
Merck
All Photos(1)

Documents

22020-U

Swagelok® Tee

Swagelok® 200-3, brass, 1/8 in. Swagelok

Synonym(s):

Swagelok® 200-3,BRASS TEE 1/8" EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
eCl@ss:
32035302
NACRES:
SB.54

material

brass

packaging

pkg of 1 ea

manufacturer/tradename

Swagelok® 200-3

fitting

1/8 in. Swagelok

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.

Application

Two Swagelok® tees connected in series maybe used in the nanoliter injection system used for microliter column chromatography, the branch of the first tee could be connected to a restricted capillary followed by high-pressure shut-off valve (“SPLIT”), whereas the branch of the second tee could be connected with wide-bore stainless steel tubing directly to a shut-off valve (“PURGE”).

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Nanoliter injection system for microcolumn liquid chromatography.
McGuffin VL and Novotny M.
Analytical Chemistry, 55 (3), 580-583 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service