Skip to Content
Merck
All Photos(1)

Key Documents

21970-U

Swagelok® 90 Degree Male Elbow

Swagelok® 200-2-2, brass, 1/8 in. Swagelok, 1/8 in. male NPT

Synonym(s):

Swagelok® 200-2-2,BRASS ELBOW MALE, 1/8TX1/8P PK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
NACRES:
SB.54

material

brass

packaging

pkg of 2 ea

manufacturer/tradename

Swagelok® 200-2-2

fitting

1/8 in. Swagelok
1/8 in. male NPT

Looking for similar products? Visit Product Comparison Guide

General description

Degree elbow is suitable to change direction of a pipe. In general, 45-degree or 90-degree elbows are used. Elbows enforce sudden inner diameter changes and rough edges which lead to turbulence and greater pressure drop. They are suitable for tubing systems. The capability to bend tubing delivers smoother transition.[1]
Nuts and ferrules included.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thwarting fluid-system leaks.
Cox, John C.
Chemical Processing (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service