Documents
20448
Ratchet wrench
wrench size 9/16 in.
About This Item
UNSPSC Code:
41100000
wrench size
9/16 in.
compatibility
for use with Swagelok 1/4 in. fittings
General description
Gets into tight places. Jaws open to slip around the nut, then close. Convenient to use in a small chromatograph oven or other confined places. 7/16in. wrench is for 1/8in. Swagelok® fittings, 9/16in. wrench is for 1/4in. Swagelok® fittings.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)