Sepabeads® SP-20SS is an epoxy-activated hydrophobic[1] styrene-divinylbenzene polymer resin.[2]
Application
Sepabeads® SP-20SS was used as adsorbent for bovine serum albumin on allyl glycidyl ether-ethylene glycol dimethacrylate copolymer using UV-visible spectrophotometry.[2]
Sepabeads® SP-20SS was used as column packing material for purification of peptides.[3]
Sepabeads® SP-20SS was used to remove active laccase from Heterobasidion annosum.[1]
Polyaromatic adsorbent resin for separation of hydrophobic compounds: small and medium proteins, peptides, amino acids. Excellent for reversed phase applications.
Other Notes
Swelling in toluene = 30%
Legal Information
Sepabeads is a registered trademark of Mitsubishi Chemical Corp.
Journal of applied microbiology, 110(4), 924-934 (2011-01-18)
Wild-type white rot fungi are the most important production organisms for laccase, a promising oxidative biocatalyst with numerous applications. This study aimed at identifying novel highly productive strains, finding optimal cultivation conditions for laccase production and establishing a simple immobilization
Applied and environmental microbiology, 77(4), 1187-1195 (2010-12-25)
Mannosylphosphorylation of N- and O-glycans, which confers negative charges on the surfaces of cells, requires the functions of both MNN4 and MNN6 in Saccharomyces cerevisiae. To identify genes relevant to mannosylphosphorylation in the dimorphic yeast Yarrowia lipolytica, the molecular functions
Allyl glycidyl ether (AGE)-ethylene glycol dimethacrylate (EGDM) copolymer with 25% crosslink density (AGE-25) shows excellent bovine serum albumin (BSA) adsorption (up to 16% (w/w)) at pH 8.0 and the adsorbed BSA is strongly bound. This protein-coated polymer provides a novel
Questions
Reviews
★★★★★ No rating value