Skip to Content
Merck
All Photos(6)

Documents

ZRB2393

Sigma-Aldrich

Anti-p-eIF4G1 (Ser1108) Antibody, clone 4G7 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Eukaryotic translation initiation factor 4 gamma 1, eIF-4-gamma 1, eIF-4G 1, eIF-4G1, p220

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 4G7
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 175 kDa
observed mol wt ~240 kDa

purified by

using Protein A

species reactivity

human, rat

species reactivity (predicted by homology)

mouse

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... EIF4G1(1981)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 4G7 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Eukaryotic translation initiation factor 4 gamma 1 (eIF4G1) phosphorylated on serine 1108.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids surrounding phosphoserine 1108 from the internal region of human Eukaryotic translation initiation factor 4 gamma 1 (eIF4G1), isoform 3.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:100 dilution of this antibody detected phospho-eIF4G1 (Ser1108) in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:100 dilution from a representative lot detected phospho-eIF4G1 (Ser1108) in lysate from Human umbilical vein endothelial cells (HUVEC).

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho-eIF4G1 (Ser1108) in PC12 cells treated with NGF.

Peptide Inhibition Assay: Target band detection in a lysate from PC12 cells treated with NGF was prevented by preblocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Eukaryotic translation initiation factor 4 gamma 1 (UniProt: Q04637-3; also known as eIF-4-gamma 1, eIF-4G 1, eIF-4G1, p220) is encoded by the EIF4G1 (also known as EIF4F, EIF4G, EIF4GI) gene (Gene ID: 1981) in human. eIF4G1 is a component of the eIF4F complex that is involved in the recognition of the mRNA cap, ATP-dependent unwinding of 5′-terminal secondary structure, and recruitment of mRNA to the ribosome. It serves as a scaffold for assembly of components of EIF4F complex that starts cap-dependent translation. This complex also plays a role in oncogenesis. Increased expression of EIF4G1 has been linked to lower median survival of patients with multiple cancers. Knockdown of eIF4G1 in prostate cancer cells is reported to reduce cyclin D1 and p-Rb protein levels and cause cell cycle delay and reduce cell viability, proliferation, and migration. eIF4G1 serves as a substrate for PKC and its phosphorylation at serine 1186 induces its binding to mitogen-activated protein kinase-interacting kinase 1 (Mnk1). Eight isoforms of eIF4G1 have been described that are produced by alternative splicing and alternative initiation. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Jaiswal, PK., et al. (2019). Cancer Cell Int. 19; Article 224; Jaiswal, PK., et al. (2018). Sci. Rep. 8; Article 7459; Dobrikov, M., et al. (2011). Mol. Cell. Biol. 31(14); 2947-2959).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service