Skip to Content
Merck
All Photos(6)

Key Documents

ZRB2340

Sigma-Aldrich

Anti-AIP Antibody, clone 1H8 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

AH receptor-interacting protein, Aryl-hydrocarbon receptor-interacting protein, HBV X-associated protein 2, Immunophilin homolog ARA9, XAP-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 1H8
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 37.66 kDa
observed mol wt ~37 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... AIP(9049)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1H8 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects AH receptor-interacting protein (AIP).

Immunogen

His-tagged, recombinant, full-length human AH receptor-interacting protein (AIP).

Application

Quality Control Testing

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected AIP in A431 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected AIP in lysates rom HepG2 cells and Human kidney tissue.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected AIP in Human kidney tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound recombinant Human AIP with a KD of 5.4 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

AH receptor-interacting protein (UniProt: O00170; also known as AIP; Aryl-hydrocarbon receptor-interacting protein, HBV X-associated protein 2, XAP-2, Immunophilin homolog ARA9) is encoded by the AIP (also known as XAP2) gene (Gene ID: 9049) in human. AIP is a highly conserved and widely distributed protein that serves as a molecular co-chaperone to heat-shock proteins. It also plays a role in aromatic hydrocarbon receptor (AHR)-mediated signaling, possibly by affecting its receptivity for ligand and/or its nuclear targeting. It contains three TPR repeats (aa 179-212; 231-264; and 265-298) in its C-terminal half that mediate many of its protein-protein interactions. It is reported to interact with RET in the pituitary gland and this interaction prevents the formation of AIP-survivin complex. In normal pituitary tissue it is detected in growth hormone and prolactin producing cells. Loss-of-function mutations in AIP gene are known to result in pituitary adenomas and somatotropinomas at early age. AIP is reported to be essential for optimal Akt signaling in response to B cell receptor stimulation. Deletion of AIP in B cells is shown to reduce Bcl6 expression and diminish adaptive immune responses. AIP protects Bcl6 from ubiquitin-mediated proteasomal degradation and helps to maintain the expression of Bcl6. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonal. (Ref.: Stojanovic, M., et al. (2019). Endocr. Connect. 8(4); 326-337; Sun, D., et al. (2019). Cell Rep. 27(5); 1461 1471).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service