Skip to Content
Merck
All Photos(11)

Key Documents

ZRB2049

Sigma-Aldrich

Anti-MMP-9 Antibody, clone 8H12 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$281.00
4 X 25 μL
$579.00

$281.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$281.00
4 X 25 μL
$579.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$281.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

8H12, recombinant monoclonal

description

8H12 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 78.46 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 8H12 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Matrix metalloproteinase-9 (MMP-9). It targets an epitope within 19 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal region of human Matrix metalloproteinase-9 (MMP-9).

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in human bone marrow tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution of this antibody detected MMP-9 in human bone marrow tissue sections.

Tested applications

Affinity Binding Assay: A representative lot of this antibody bound MMP-9 with a KD of 8.3 x 10-9 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected MMP-9 in THP-1 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-MMP-9, clone 8H12 ZooMAb®, Cat. No. ZRB2049, is a recombinant Rabbit monoclonal antibody that targets MMP-9 and is tested for use in Affinity Binding Assay, Immunocytochemistry, and Immunohistochemistry (Paraffin).

Target description

Matrix metalloproteinase-9 (UniProt: P14780; also known as MMP-9, 92 kDa gelatinase, 92 kDa type IV collagenase, Gelatinase B, GELB) is encoded by the MMP9 (also known as CLG4B) gene (Gene ID: 4318) in human. MMP-9 is an extracellular or secreted enzyme that is synthesized as inactive proenzyme with a signal peptide (aa 1-19) and an activation peptide (aa 23-93) that are cleaved to generate the active enzyme. It is a zinc-binding enzyme that binds two zinc ions per subunit. MMP-9 contains a conserved cysteine that is present in the cysteine-switch motif and binds the catalytic zinc ion, thus inhibiting the enzyme. The dissociation of the cysteine from the zinc ion upon the activation-peptide release activates the enzyme. Processing of the precursor is reported to yield different active forms of 64, 67 and 82 kDa. It can be present as a monomer or a disulfide-linked homodimer. MMP-9 is produced by normal alveolar macrophages and granulocytes. Macrophages and transformed cells usually produce only the monomeric form. MMP-9 plays an essential role in local proteolysis of the extracellular matrix, in leukocyte migration, and bone osteoclastic resorption. It cleaves type IV and type V collagen into large C-terminal three quarter fragments and shorter N-terminal one quarter fragments. MMP-9 expression is regulated by several cytokines and growth factors. In coordination with other MMPs, it plays a role in normal tissue remodeling events, including neurite growth, embryonic development, angiogenesis, ovulation, mammary gland involution and wound healing. Abnormalities in MMP-9 are associated with a variety of autoimmune diseases, including systemic lupus erythematosus, Sjogren′s syndrome, systemic sclerosis, rheumatoid arthritis, multiple sclerosis, polymyositis and atherosclerosis. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service