ZRB2047
Anti-CBX1 Antibody, clone 1D7 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
Chromobox protein homolog 1;HP1Hsbeta;Heterochromatin protein 1 homolog beta;HP1 beta;Heterochromatin protein p25;M31;Modifier 1 protein;p25beta
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
1D7, recombinant monoclonal
description
1D7 Clone
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 21.42 kDa
observed mol wt ~22 kDa
purified by
using Protein A
species reactivity
rat, mouse, human
species reactivity (predicted by homology)
canine, monkey, porcine, feline, bovine
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
C-terminal
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... CBX1(10951)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in Caco-2 cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CBX1 in Caco-2 cell lysate.
Tested Applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CBX1 in HeLa, L6, and NIH3T3 cell lysates.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CBX1 in A431, HepG2, and NIH3T3 cells.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CBX1 in human testis tissue sections.
Affinity Binding Assay: A representative lot of this antibody bound CBX1 peptide with a KD of 3.1 x 10-7 in an affinity binding assay.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service