Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1977

Sigma-Aldrich

Anti-ERK5 Antibody, clone 2H15 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

BMK-1, Big MAP kinase 1, EC:2.7.11.24, Extracellular signal-regulated kinase 5, MAP kinase 7, MAPK 7, Mitogen-activated protein kinase 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2H15, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

human, mouse

species reactivity (predicted by homology)

monkey, porcine, canine, bovine, feline

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminus

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2H15 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects ERK5. It targets an epitope within 18 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminal region of human ERK5.

Application

Quality Control Testing

Evaluated by Western Blotting in NIH3T3 cell nuclear extract.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected ERK5 in NIH3T3 cell nuclear extract.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution of this antibody detected ERK5 in human spleen tissue sections.

Affinity Binding Assay:: A representative lot of this antibody bound ERK5 peptide with a KD of 1.3 x 10-8 in an affinity binding assay.

Enzyme Immunoassay (ELISA) Analysis: A serial of dilutions from a representative lot detected recombinant Human ERK5.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected GST-tagged recombinant Human ERK5.

Flow Cytometry Analysis: 0.1 µg from a representative lot detected ERK5 in one million A549 and HeLa cells.

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected ERK5 in Human spleen tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Mitogen-activated protein kinase 7 (UniProt: Q13164; also known as EC:2.7.11.24, MAP kinase 7, MAPK 7, Big MAP kinase 1, BMK-1, Extracellular signal-regulated kinase 5, ERK-5) is encoded by the MAPK7 (also known as BMK1, ERK5, PRKM7) gene (Gene ID: 5598) in human. ERK5 is an unusual protein kinase with a kinase domain (aa 55-347) and a transcriptional transactivation domain (aa 664-789). It is expressed in many adult tissues and higher expression is observed in heart, placenta, lung, kidney, and skeletal muscle. Although its kinase domain  displays high sequence identity with ERK1/2, they tend to have distinct substrates and unlike ERK1/2, it has a large C-terminal extension that contains a NuCLEAR localization signal (aa 505-539) and the transcriptional transactivation domain. ERK5 plays a role in various cellular processes, including cell proliferation, differentiation, and cell survival. Its upstream activator is MAP2K5 and following activation, it translocates to the nucleus and phosphorylates various downstream targets. It is activated by Dual® phosphorylation of threonine 219 and tyrosine 221. ERK5 is also important for endothelial function and maintenance of blood vessel integrity. ERK5-deficient mice die around embryonic day 10.5 because of cardiovascular defects and angiogenic failure in embryonic and extraembryonic tissues.  Four isoforms of ERK5 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: (Ref.: Cook, SJ., and Lochhead, PA. (2022). Front. Cell Dev. Biol. 10; 839997; Yang, Q., et al. (2013). Oncogene. 32(26); 3156-3264; Nishimoto, S., and Nishida, E.  (2006). EMBO Rep. 7(8); 782-786).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
Dual is a registered trademark of Ciba-Geigy AG
NuCLEAR is a trademark of Sigma-Aldrich Co. LLC
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service