Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1848

Sigma-Aldrich

Anti-Cdc42 Antibody, clone 2D16 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$281.00
4 X 25 μL
$579.00

$281.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$281.00
4 X 25 μL
$579.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$281.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2D16, recombinant monoclonal

description

2D16 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 21.26 kDa
observed mol wt ~20 kDa

purified by

using Protein A

species reactivity

mouse, human, rat

species reactivity (predicted by homology)

feline, zebrafish, canine, bovine, monkey, porcine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CDC42(998)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2D16 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Cell division control protein 42 homolog (Cdc42). It targets an epitope within 19 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal half of human Cell division control protein 42 homolog (Cdc42).

Application

Quality Control Testing

Evaluated by Western Blotting in NIH3T3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Cdc42 in NIH3T3 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Cdc42 in U2OS, HEK293, and C6 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected CDC42 in human kidney tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Cdc42 in U2OS cells.

Affinity Binding Assay: A representative lot of this antibody bound Cdc42 with a KD of 3.0 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Cell division control protein 42 homolog (UniProt: P60953; also known as EC:3.6.5.2, G25K GTP-binding protein, CDC42) is encoded by the CDC42 gene (Gene ID: 998) in human. Cdc42 is a plasma membrane-associated small GTPase that cycles between an active GTP-bound and an inactive GDP-bound state. In its active state it binds to a variety of effector proteins to regulate cellular responses. Its activity is regulated by guanine nucleotide exchange factors (GEFs), which promote the exchange of bound GDP for free GTP, GTPase activating proteins (GAPs) that increase the GTP hydrolysis activity, and GDP dissociation inhibitors, which inhibit the dissociation of the nucleotide from the GTPase. Cdc42 is known to regulate the dynamic organization of the cytoskeleton and membrane trafficking for physiologic processes such as cell proliferation, motility, polarity, cytokinesis, and growth. It is involved in epithelial cell polarization processes and regulates the bipolar attachment of spindle microtubules to kinetochores before chromosome congression in metaphase. During the prometaphase stage, it localizes to spindle and moves to the central spindle as cells progress through anaphase to telophase. Cdc42 is also reported to play a role in phagocytosis through organization of the F-actin cytoskeleton associated with forming phagocytic cups. Deregulation of Cdc42 is found in several pathogenic processes such as cancer, cardiovascular diseases, and neuronal degenerative diseases. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Schlam, D., et al. (2015). Nat. Commun. 6; 8623; Oceguera-Yanez, F., et al. (2005). J. Cell Biol. 168(2); 221-232).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service