Skip to Content
Merck
All Photos(9)

Documents

ZRB1820

Sigma-Aldrich

Anti-IQGAP1 Antibody, clone 4M20 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4M20, recombinant monoclonal

description

4M20 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 189.25 kDa
observed mol wt ~190 kDa

purified by

using Protein A

species reactivity

hamster, monkey, human

species reactivity (predicted by homology)

mouse, porcine, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... IQGAP1(8826)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 4M20 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Ras GTPase-activating-like protein IQGAP1. It targets an epitope within 18 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminal region of human Ras GTPase-activating-like protein IQGAP1.

Application

Quality Control Testing

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected IQGAP1 in HEK293 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected IQGAP1 in CHO K1, COS-7, and A549 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected IQGAP1 in human colon tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound IQGAP1 with a KD of 5.6 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Ras GTPase-activating-like protein IQGAP1 (UniProt: P46940; also known as p195) is encoded by the IQGAP1 (also known as KIAA0051) gene (Gene ID: 8826) in human. IQGAP1 is a cell-cell contact regulator that also plays a crucial role in regulating the dynamics and assembly of the actin cytoskeleton. Its expression is reported in the placenta, lung, and kidney and low level of expression is observed in the heart, liver, skeletal muscle, and pancreas. In eukaryotic cells IQGAP1 binds to and alters the function of several proteins, including actin, E-cadherin, beta-catenin, Cdc42, and Rac1. It is localized in the nucleus, cytoplasm, and membrane and its subcellular distribution is regulated by the cell cycle. Its nuclear levels are reported to increase at the G1/S phase. It may also play a role in cell cycle regulation by contributing to cell cycle progression after DNA replication arrest. IQGAP1 contains a calponin homology domain (aa 44-159 that is responsible for actin binding, a WW motif (aa 679-712) that is essential for the association of ERK2, four calmodulin-binding IQ domains (aa 745-864), and a RasGAP-related domain (aa 1004-1237) that binds the small GTPases Cdc42 and Rac1. IQGAP1 is reported to inhibit GTPase activity of Cdc42 and overexpression of IQGAP1 in cells is shown to increase Cdc42-GTP levels. Two short C-terminal fragments of IQGAP1 (C1: aa 956-1274; and C2: 1276-1657) are shown to bind exclusively to nucleotide-depleted Cdc42 (Cdc42-ND), whereas the entire C-terminus (aa 956-1657) binds predominantly to the Cdc42-GTP S. Phosphorylation of IQGAP1 at Ser 1443 by protein kinase Cϵ is reported to inhibit the intramolecular interaction of the C-terminal fragments. Ser 1443 phosphorylation causes an opening of the IQGAP1 structure and exposes C1 and C2 regions. Ser 1443 phosphorylation also enhances the ability to promote neurite outgrowth. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Li, Z., et al. (2005). J. Biol. Chem. 280(14);13871-13878; Grohmanova, K., et al. (2004). J. Biol. Chem. 279(47); 48495-48504).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service