Skip to Content
Merck
All Photos(9)

Key Documents

ZRB1813

Sigma-Aldrich

Anti-G0S2 Antibody, clone 2C13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$281.00
4 X 25 μL
$579.00

$281.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$281.00
4 X 25 μL
$579.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$281.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2C13, monoclonal
recombinant monoclonal

description

2C13 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~11.32 kDa
observed mol wt ~8 kDa

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2C13 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects G0/G1 switch protein 2 (G0S2).

Immunogen

GST/His-tagged full-length recombinant human G0/G1 switch protein 2 (G0S2).

Application

Quality Control Testing

Evaluated by Western Blotting in Rat Heart tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected G0S2 in Rat Heart tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected G0S2 in lysates from human heart, mouse heart, and A7R5 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected G0S2 in rat heart tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound G0S2 protein with a KD of 2.5 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-G0S2, clone 2C13 ZooMAb®, Cat. No. ZRB1813, is a recombinant Rabbit monoclonal antibody that specifically targets G0S2 and is tested for use in Affinity Binding Assay, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

G0/G1 switch protein 2 (UniProt: P27469; also known as G0/G1 switch regulatory protein 2, Putative lymphocyte G0/G1 switch gene, G0S2) is encoded by the G0S2 gene (Gene ID: 50486) in human. G0S2 is a highly conserved, mitochondrial protein that is widely expressed in vertebrate species. High expression of G0S2 is reported in peripheral blood cells, skeletal muscle, heart, liver and kidney. It is multifaceted protein that is involved in cell proliferation, apoptosis, inflammation, metabolism, and carcinogenesis. The G0S2 gene is reported to be hypermethylated in several human cancer cell lines. Its promoter region contains potential binding sites for AP1, AP2, AP3, and Nuclear factor of activated T cells (NFAT). The G0S2 protein is shown to contain two -helices that are separated by a hydrophobic sequence with the potential to generate turns and assume a β-sheet conformation. It also has several putative phosphorylation sites for protein kinase C (PKC) and casein kinase II. G0S2 can interact with Bcl2 and block the formation of Bcl2-Bax heterodimer to block apoptosis. In fat tissue it acts as a molecular brake on triglyceride catabolism. G0S2 is reported to be a major selective inhibitor of adipose triglyceride lipase (AGTL) and its hydrolase activity. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Heckmann, BL., et al. (2013). Biochim. Biophys. Acta. 1831(2); 276-281; Heckmann, BL., et al. (2016). PLoS ONE 11(6); e0156742).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service