Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1809

Sigma-Aldrich

Anti-COIL Antibody, clone 2K14-2 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

p80-coilin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2K14-2, recombinant monoclonal

description

2K14-2 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 62.61 kDa
observed mol wt ~Target molecular weight 38 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... COIL(8161)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2K14-2 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Coilin (COIL). It targets an epitope within 25 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the C-terminal region of human Coilin (COIL).

Application

Quality Control Testing

Evaluated by Western Blotting with His-ABP-tagged recombinant fragment of Human Coilin (COIL).

Western Blotting Analysis (WB): A 1:10,000 dilution of this antibody detected His-ABP tagged recombinant fragment of Human Coilin (COIL).

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Coilin (COIL) in HepG2 cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution of this antibody detected Coilin (COIL) in Human testis tissue sections.

Affinity Binding Assay:: A representative lot of this antibody bound Coilin peptide with a KD of 3.0 x 10-6 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected COIL in HepG2 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Coilin (UniProt: P38432; also known as p80-coilin) is encoded by the COIL (also known as CLN80) gene (Gene ID: 8161) in human. Coilin is a nuclear phosphoprotein that is express in all cell types. It is a component of nuclear coiled bodies (Cajal bodies) that are involved in the modification and assembly of nucleoplasmic snRNPs. It is shown to be hyperphosphorylated during mitosis and this hyperphosphorylation triggers Cajal bodies disassembly during cell replication. Sixteen different phosphorylated residues have been identified in coilin, five of these are found primarily in only mitotic cell preparations and eight in only interphase samples. Its first N-terminal 92 amino acids are essential for self-association leading to its characteristic fibrous pattern observed in Cajal bodies. In the central region it has two nuclear localization signals and a nucleolar localization sequence. The conserved C-terminal region contains an atypical Tudor domain (aa 460-559) that contains four characteristic β-strands (β1β2 and β3β4) and one short 310 helix. The final ten residues at the C-terminal region have been implicated in controlling availability of the N-terminus for self-interaction, Cajal bodies formation, and coilin localization. Symmetrical dimethylation of arginine residues within the RG (arginine-glycine) repeat region (aa 423-420) of coilin is reported to enhance its affinity for SMN, and thus localization of SMN complexes to Cajal bodies. Coilin is suggested to promote RNP biogenesis by acting as a chaperone of nuclear small non-coding RNAs, which might not be essential in differentiated cells, but may become critical in rapidly developing embryos. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Machyna, M., et al. (2015). RNA Biol. 12(6); 590-596; Shanbhag, R., et al. (2010). FEBS Lett. 584(20); 4351-4356; Toyota, CG., et al. (2010). Chromosoma. 119(2); 205-215; Carmo-Fonseca, M., et al. (1993). J. Cell Biol. 120(4); 841-852).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

0.3 mg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service