Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1726

Sigma-Aldrich

Anti-HSC70 Antibody, clone 1B15 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Heat shock cognate 71 kDa protein;EC:3.6.4.10;Heat shock 70 kDa protein 8;Lipopolysaccharide-associated protein 1;LAP-1;LPS-associated protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$281.00
4 X 25 μL
$579.00

$281.00


Estimated to ship onApril 26, 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$281.00
4 X 25 μL
$579.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$281.00


Estimated to ship onApril 26, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B15, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 70.9 kDa
observed mol wt ~70 kDa

purified by

using Protein A

species reactivity

human, rat, mouse

species reactivity (predicted by homology)

bovine, hamster, monkey, feline, canine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... HSPA8(3312)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1B15 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects HSC70. It targets an epitope within 20 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of human HSC70.

Application

Quality Control Testing

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected HSC70 in A431 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected HSC70 in lysates from L6, HL-60 cells and mouse heart tissue.

Affinity Binding Assay: A representative lot of this antibody bound in HSC70 with a KD of 2.9 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected HSC70 in Caco-2 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected HSC70 in human testis tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Heat shock cognate 71 kDa protein (UniProt: P11142; also known as EC:3.6.4.10, Heat shock 70 kDa protein 8, Lipopolysaccharide-associated protein 1, LAP-1, LPS-associated protein 1, HSC70) is encoded by the HSPA8 (also known as HSC70, HSP73, HSPA10) gene (Gene ID: 3312) in human. HSC70 is a ubiquitously expressed protein that serves as a molecular chaperone and is implicated in variety of cellular processes, including protection from the proteome stress, folding and transport of newly synthesized polypeptides, activation of proteolysis of misfolded proteins. Hence, it plays a vital role in protein quality control system to ensure proper folding and refolding of misfolded proteins. It also plays a critical role in mitochondrial import and delivers preproteins to the mitochondrial import receptor TOMM70. These functions of HSC70 are achieved through cycles of ATP binding, ATP hydrolysis and ADP release, mediated by co-chaperones. The co-chaperones not only regulate different steps of the ATPase cycle of HSC70, but they also have an individual specificity such that one co-chaperone may promote folding of a substrate while another may promote degradation. Its N-terminal nucleotide binding domain (NBD; aa 2-386) is responsible for binding and hydrolyzing ATP and the C-terminal substrate-binding domain (SBD; aa 394-509) binds to the client/substrate proteins. The two domains are allosterically coupled so that, when ATP is bound to the NBD, the SBD binds relatively weakly to client/substrates. When ADP is bound in the NBD, a conformational change enhances the affinity of the SBD for client/substrate proteins. HSC70 is mainly localized in cytoplasmic mRNP granules containing untranslated mRNAs. However, it translocates rapidly from the cytoplasm to the nuclei upon heat shock. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Young, JC., et al. (2003). Cell. 112(1); 41-50; Takayama, S., et al. (1997). EMBO J. 16(16); 4887-4896).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service