Skip to Content
Merck
All Photos(16)

Key Documents

ZRB1708

Sigma-Aldrich

Anti-IL-17A Antibody, clone 1F21 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Interleukin-17A;IL-17;Cytotoxic T-lymphocyte-associated antigen 8;CTLA-8

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$281.00
4 X 25 μL
$579.00

$281.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$281.00
4 X 25 μL
$579.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$281.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F21, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 17.5 kDa
observed mol wt ~15 kDa

purified by

using Protein A

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... IL17A(3605)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone IF21 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects IL-17A. It targets an epitope within 21 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the internal region of human Interleukin-17A (IL-17A).

Application

Quality Control Testing

Evaluated by Western Blotting in Mouse Thymus tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected IL-17A in Mouse Thymus tissue lysate.

Tested applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected IL-17A in HL-60 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected IL-17A in human colon and human thymus tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound IL-17A with a KD of 3.4 x 10-8 in an affinity binding assay.

Flow Cytometry Analysis: 1 μg from a representative lot detected IL-17A in one million HL-60 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Interleukin-17A (UniProt: Q16552; also known as IL-17, IL-17A, Cytotoxic T-lymphocyte-associated antigen 8, CTLA-8) is encoded by the IL17A (also known as CTLA8, IL17) gene (Gene ID: 3605) in human. IL-17A is a proinflammatory cytokine secreted by activated CD4+ Th17 cells. It is synthesized with a signal peptide (aa 1-23) that is subsequently cleaved off to produce the mature protein. Its levels are up-regulated by IL23A-IL12B. It is an effector cytokine of innate and adaptive immune system involved in antimicrobial host defense and maintenance of tissue integrity. It plays important roles in multiple biological processes, including the induction of IL-6, IL-8, G-CSF, and PGE2 production in epithelial, endothelial or fibroblasts; the enhancement of surface expression of ICAM-1 in fibroblasts; activation of NF-kB and co-stimulation of T cell proliferation. It is also involved in antimicrobial host defense and maintenance of tissue integrity. Its actions are mediated via interaction with IL-17RA and IL-17RC heterodimeric receptor complex, triggering homotypic interaction of these receptors with TRAF3 Interacting Protein 2 (TRAF3IP2) adapter. This leads to downstream TRAF6-mediated activation of NF-kB and MAP kinase pathways resulting in transcriptional activation of cytokines, chemokines, antimicrobial peptides, and matrix metalloproteinases. IL-17A also plays an important role in connecting T cell-mediated adaptive immunity and acute inflammatory response to destroy extracellular bacteria and fungi. In synergy with IL-17F, it is shown to mediate the production of antimicrobial b-defensins DEFB1, DEFB103A, and DEFB104A by mucosal epithelial cells, which limits the entry of microbes through the epithelial barriers. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Boisson, B., et al. (2013). Immunity. 39(4); 676-686; Wright, J.F., et al. (2008). J. Immunol. 181(4); 2799-2805).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service