Skip to Content
Merck
All Photos(9)

Key Documents

ZRB1660

Sigma-Aldrich

Anti-RhoB Antibody, clone 3M19 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Rho-related GTP-binding protein RhoB

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$281.00
4 X 25 μL
$579.00

$281.00


Estimated to ship onApril 19, 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$281.00
4 X 25 μL
$579.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$281.00


Estimated to ship onApril 19, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3M19, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 22.12 kDa
observed mol wt ~22 kDa

purified by

using Protein A

species reactivity

human, rat

species reactivity (predicted by homology)

canine, mouse, bovine, monkey, porcine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... RHOB(388)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3M19 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects RhoB. It targets an epitope within 17 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal half of human RhoB.

Application

Quality Control Testing

Evaluated by Western Blotting in MCF7 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected RhoB in MCF7 cell lysate.

Tested applications


Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected RhoB in HEK293 cells and 1:10,000 dilution detected RhoB in Rat Brain tissue lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected RhoB in human cerebral cortex tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound RhoB with a KD of 4.6 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Rho-related GTP-binding protein RhoB (UniProt: P62745; also known as Rho cDNA clone 6, h6) is encoded by the RHOB (also known as ARH6, ARHB) gene (Gene ID: 388) in human. RhoB is a member of the Ras super-family of small GTP-binding proteins that is synthesized with a short propeptide (aa 194-196), which is subsequently cleaved off to produce the mature active form. It has three nucleotide-binding regions (aa 12-19; 59-63; and 117-120). It is involved in hydrolysis of GTP and is active in GTP-bound form. RhoB is a short-lived Rho GTPase whose expression is inducible by a variety of stimuli including growth factors and stress stimuli. Its levels are up-regulated by DNA damaging agents, such as hydrogen peroxide or ionizing radiation. It plays a negative role in tumorigenesis as its deletion is known to cause tumor formation. It is reported to mediate apoptosis in neoplastically transformed cells following DNA damage. It is not considered to be essential for development but affects cell adhesion and growth factor signaling in transformed cells. RhoB participates in sorting and degradation of growth factors and cytokine receptors. It is shown to be critical for the inflammatory response of endothelial cells to TNFa, possibly through MAP kinase activation downstream of the TNFR. RhoB displays 83% homology with RhoA. However, unlike RhoA, which is cytosolic and translocates to plasma membrane upon activation, RhoB localizes to endosomes/multivesicular bodies. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Vega, FM., and Ridley, AJ. (2018). Small GTPases. 9(5); 384-393).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service