Skip to Content
Merck
All Photos(20)

Key Documents

ZRB1542

Sigma-Aldrich

Anti-phospho-p53 (Ser15) Antibody, clone 5L22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$281.00
4 X 25 μL
$579.00

$281.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$281.00
4 X 25 μL
$579.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$281.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5L22, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 43.65 kDa
observed mol wt ~51 kDa

purified by

using Protein A

species reactivity

monkey, human

species reactivity (predicted by homology)

porcine, bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer15)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 5L22 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects p53 protein phosphorylated on serine 15.

Immunogen

KLH-conjugated linear peptide corresponding to 11 amino acids surrounding phosphoserine 15 from the N-terminal region of human p53.

Application

Quality Control Testing

Evaluated by Western blotting with recombinant fragment of human p53 surrounding phosphoserine 15.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected recombinant human p53 fragment phosphorylated on serine 15 but did not react with non-phosphorylated construct (p53 construct: Courtesy of Dr. Jesse Rinehart, Yale University, School of Medicine).

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected phospho-p53 (Ser15) in lysate from Cos-1 cells treated with UV light.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected phospho-p53 (Ser15) in one million A549 cells treated with UV light

Affinity Binding Assay: A representative lot of this antibody bound phospho-p53 (Ser15) with a KD value of 9.7 x 10-8 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected phospho-p53 (Ser15) in human breast cancer and human colon cancer tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho-p53 (Ser15) in Cos-1 cells treated with UV light..

Peptide Inhibition Assay: Target band detection in lysate from Cos-1 cells treated with UV light was prevented by pre-blocking of a representative lot with the p53 phospho-peptide (Ser15), but not the corresponding nonphosphopeptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
This monoclonal Anti-phospho-p53 (Ser15), clone 5L22 ZooMAb® Rabbit Monoclonal, Cat. No. ZRB1542 is used in Western Blotting, Peptide Inhibition, Immunocytochemistry, Immunohistochemistry, Affinity Binding Assay, Flow Cytometry, for phospho-p53 (Ser15).

Target description

Cellular tumor antigen p53 (UniProt: P04637; also known as Antigen NY-CO-13, Phosphoprotein p53, Tumor suppressor p53) is encoded by the TP53 (also known as P53) gene (Gene ID: 7157) in human. p53, a well-conserved phosphoprotein, is one of the best-known tumor suppressors. Human p53 consists of 393 amino acids assembled into five structurally and functionally different domains. It acts as a sequence-specific nuclear transcription factor that binds to defined consensus sites within DNA as a tetramer and represses transcription of a set of genes involved in cell growth stimulation, while activating a different set of genes involved in cell cycle control. It causes growth arrest before either DNA replication thereby providing a window for DNA repair or elimination of cells with severely damaged DNA strands. In some cell types, however, p53 activation results in apoptosis as a means of eliminating severely damaged cells. In normal, non-activated cells, the p53 signaling network is not active. However, p53 signaling is activated in cells as a response to various signals. Higher levels of p53 are found in a wide variety of transformed cells and it is mutated or inactivated in about 60% of cancers. Carcinogen-induced DNA damage, abnormal proliferative signals, hypoxia, and loss of cell adhesion are some of the most common signals that activate p53. The final outcome of p53 activation depends on many factors and is mediated largely through the action of downstream effector genes transactivated by p53. In unstressed cells p53 is latent and is maintained at low levels by targeted, ubiquitin-mediated degradation mediated by MDM2 and many other ubiquitin ligases. MDM2 functions as an E3 ligase that ubiquitinates p53 and forces its export from the nucleus to the cytoplasm, where p53 is degraded by the proteasome. p53 is shown to undergo several post-translational modifications that affect its activity. p53 acetylation on lysine 382 enhances its transcriptional activity. Site specific phosphorylations can affect its activity differently. Phosphorylation at serine 9 by HIPK4 increases its repression activity on BIRCS promoter. It can undergo phosphorylation on serine 15 upon UV treatment, which is enhanced by interaction with BTG3 Associated Nuclear Protein (BANP). Phosphorylation on serine 20 by Check 2 in response to DNA damage prevents its ubiquitination by MDM2. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service