Skip to Content
Merck
All Photos(10)

Key Documents

ZRB1471

Sigma-Aldrich

Anti-TRPA1 Antibody, clone 2N14 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$281.00
4 X 25 μL
$579.00

$281.00


Estimated to ship onApril 12, 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$281.00
4 X 25 μL
$579.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$281.00


Estimated to ship onApril 12, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2N14, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 127.5 kDa
observed mol wt ~40 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal cytoplasmic domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TRPA1(8989)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2N14 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Transient receptor potential cation channel subfamily A member 1 (TRPA1). It targets an epitope within 18 amino acids from the N-terminal, cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the N-terminal, cytoplasmic domain of human Transient receptor potential cation channel subfamily A member 1 (TRPA1)

Application

Quality Control Testing

Evaluated by Western Blotting with His-tagged full-length recombinant Human TRPA1 protein.

Western Blotting Analysis (WB): A 1:10,000 dilution of this antibody detected His-tagged recombinant fragment of Human TRPA1 protein.

Tested applications

Enzyme Immunoassay Analysis (ELISA): A representative lot of this antibody detected His-tagged recombinant fragment of TRPA1 in ELISA applications.

Affinity Binding Assay:: A representative lot of this antibody bound TRPA1 peptide with a KD of 1.0 x 10-12 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TRPA1 in SH-SY5Y cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Transient receptor potential cation channel subfamily A member 1 (UniProt: O75762; also known as Ankyrin-like with transmembrane domains protein 1, Transformation-sensitive protein p120, Wasabi receptor) is encoded by the TRPA1 (also known as ANKTM1) gene (Gene ID: 8989) in human. TRPA1 is a multi-pass membrane protein with four cytoplasmic domains, six transmembrane domains, four cytoplasmic domains, and one pore-forming regions (aa 902-933). It contains 14 N-terminal ankyrin repeats and serves as a receptor-activated non-selective cation channel involved in pain detection and perception of cold, itch, and oxygen concentration perception. It displays about 8-fold greater preference for divalent over monovalent cations. It can be activated at sites of tissue injury or sites of inflammation directly by endogenous mediators or indirectly as a downstream target via signal transduction from a number of distinct GPCRs. One of the specific functions of TRPA1 involves the detection, integration, and initiation of pain signals in the peripheral nervous system. Is activated by a large variety of structurally unrelated electrophilic and non-electrophilic chemical compounds. Electrophilic ligands are reported to activate TRPA1 by interacting with critical N-terminal Cys residues in a covalent manner. Under normoxia, hydroxylation of proline residue in the N-terminal region is required for inhibition of TRPA1 activity. In hypoxic conditions, a decrease in oxygen concentration diminishes the activity of the HIF-prolyl hydroxylase 2 (EGLN1) and relieves TRPA1 inhibition leading to channel activation. However, in oxidation of Cys-633 and Cys-856 in hyperoxia can override the hydroxylase EGLN1-mediated inhibition, causing TRPA1 activation. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Miyake, T., et al. (2016). Nat. Commun. 7; Article 12840; Moparthi, L., et al. (2014). Proc. Natl. Acad. Sci. USA. 111(47); 16901-16906).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service