Skip to Content
Merck
All Photos(11)

Key Documents

ZRB1453

Sigma-Aldrich

Anti-Bak Antibody, clone 2D18 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Apoptosis regulator BAK, Bcl-2 homologous antagonist/killer, Bcl-2-like protein 7, Bcl2-L-7

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$239.00
4 X 25 μL
$492.00

$239.00


Estimated to ship onApril 11, 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$239.00
4 X 25 μL
$492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$239.00


Estimated to ship onApril 11, 2025


Request a Bulk Order

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2D18, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 23.41 kDa
observed mol wt 23 kDa

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... BAK1(578)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2D18 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Bcl-2 homologous antagonist/killer (BAK). It targets an epitope within 16 amino acids from the N-terminal region.

Immunogen

KLH conjugated linear peptide corresponding to 16 amino acids from the N-terminal region of human Bcl-2 homologous antagonist/killer (BAK).

Application

Anti-Bak, clone 2D18 ZooMAb, Cat. No. ZRB1453, is a recombinant Rabbit monoclonal antibody that specifically detects Bak and is tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Bak in RAW264.7 cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Bak in human urinary bladder tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Bak in Jurkat cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Bcl-2 homologous antagonist/killer (UniProt: Q16611; also known as Apoptosis regulator BAK, Bcl-2-like protein 7, Bcl2-L-7, BAK) is encoded by the BAK1 (also known as BAK, BCL2L7, CDN1) gene (Gene ID: 578) in human. BAK is a single-pass membrane protein that plays a role in the mitochondrial apoptosis process. It is homodimeric protein whose homodimerization is zinc-dependent. It can also form heterodimers with BCL2 and BCL2L1. It is expressed in a wide variety of tissues, with highest levels displayed in heart and skeletal muscle. In non-apoptotic cells, BAK constitutively resides in the mitochondrial outer membrane, but upon arrival of cell death signals, it promotes mitochondrial outer membrane (MOM) permeabilization by oligomerizing to form pores within the MOM and releases apoptogenic factors into the cytosol, including cytochrome c, promoting the activation of caspase 9. BAK contains three BH motifs (BH1: aa 117-136; BH2: aa 169-184; and BH3: aa 74-88) and intact BH3 motif is required by BIK, BID, BAD, and BAX for their pro-apoptotic activity and their interaction with anti-apoptotic members of the Bcl-2 family. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

0.3 mg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service