Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1444

Sigma-Aldrich

Anti-PEA15 Antibody, clone 2J13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

15 kDa phosphoprotein enriched in astrocytes, Astrocytic phosphoprotein PEA-15, PED, Phosphoprotein enriched in diabetes

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$211.00
4 X 25 μL
$434.00

$211.00


Estimated to ship onMarch 28, 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$211.00
4 X 25 μL
$434.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ICC
IHC (p)
WB
affinity binding assay
clone:
2J13, monoclonal
recombinant monoclonal
species reactivity:
rat, mouse, human, monkey
citations:
technique(s):
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

$211.00


Estimated to ship onMarch 28, 2025


Request a Bulk Order

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2J13, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 15.04 kDa
observed mol wt 15 kDa

species reactivity

rat, mouse, human, monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PEA15(8682)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2J13 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Astrocytic phosphoprotein PEA-15.

Immunogen

GST/His-tagged full-length recombinant human Astrocytic phosphoprotein PEA-15.

Application

Anti-PEA15, clone 2J13 ZooMAb, Cat. No. ZRB1444, is a recombinant Rabbit monoclonal antibody that specifically targets PEA15 and is tested in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PEA15 in NIH3T3 and C6 cell lysates.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PEA15 in NIH 3T3 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected PEA15 in human cerebral cortex tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound recombinant PEA15 protein with a KD of 2.7 x 10-5 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Astrocytic phosphoprotein PEA-15 (UniProt: Q15121; alsoknown as 15 kDa phosphoprotein enriched in astrocytes, Phosphoprotein enrichedin diabetes, PED) is encoded by the PEA15 gene (Gene ID: 8682) in human.PEA-15is a cytoplasmic protein that is ubiquitously expressed. Higher expressions arefound in ear, brain, muscle, and adipose tissue. Higher levels of expressionhave been reported in tissues from individuals with type 2 diabetes. PEA-15blocks Ras-mediated inhibition of integrin activation and modulates the ERK MAPkinase cascade. It is reported to Inhibit RPS6KA3 activities by retaining it inthe cytoplasm. It is also shown to inhibits both TNFRSF6- and TNFRSF1A-mediatedcaspase 8 activity and apoptosis. It also regulates glucose transport by controllingboth the content of SLC2A1 glucose transporters on the plasma membrane and theinsulin-dependent trafficking of SLC2A4 from the cell interior to the surface.PEA-15 is phosphorylated by protein kinase C and calcium-calmodulin-dependentprotein kinase. These phosphorylation events are modulated by neurotransmittersor hormones. PEA-15 has two microtubule binding sites (aa 98-107 and122-129). This ZooMAb® recombinantmonoclonal antibody, generated by our proprietary technology, offerssignificantly enhanced specificity, affinity, reproducibility, and stabilityover conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in lyophilized from PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

0.3 mg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service