Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1287

Sigma-Aldrich

Anti-CD11c Antibody, clone 1D6 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$281.00
4 X 25 μL
$579.00

$281.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$281.00
4 X 25 μL
$579.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$281.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D6, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 127.38 kDa
observed mol wt ~130 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1D6 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects CD11c. It targets an epitope within 15 amino acids form the N-terminal, extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the N-terminal, extracellular domain of human Integrin alpha-X (CD11c).

Application

Quality Control Testing

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CD11c in THP-1 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CD11c in U937 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CD11c in THP-1 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CD11c in human tonsil tissue sections.

Flow Cytometry Analysis: 1 μg from a representative lot detected CD11c in one million THP-1 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-CD11c, clone 1D6 ZooMAb®, Cat. No. ZRB1287, is a recombinant Rabbit monoclonal antibody that specifically targets CD11c and is tested for use in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Integrin alpha-X (UniProt: P20702; also known as CD11 antigen-like family member C, Leu M5, Leukocyte adhesion glycoprotein p150,95 alpha chain, Leukocyte adhesion receptor p150-95, CD11c) is encoded by the ITGAX (also known as CD11C) gene (Gene ID: 3687) in human. CD11c is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off to generate the mature form that contains a long extracellular domain (aa 20-1107), a transmembrane domain (aa 1108-1128), and a cytoplasmic domain (aa 1129-1163). CD11c is predominantly expressed in macrophages, monocytes, and granulocytes and along with integrin b2 it serves as a receptor for fibrinogen. It recognizes the sequence G-P-R in fibrinogen. The aXb2 heterodimer binds many ligands, including collagen type 1, iC3b, fibrinogen, ICAM-1, and lipopolysaccharide. It is reported to mediate cell-cell interaction during inflammatory responses and is especially important in monocyte adhesion and chemotaxis. The I domains of CD11 are responsible for the binding of ligands and have a unique structure with 6-7 a -helixes and 6 b sheets with interconnecting loops. In CD11c the loops on the I domain are involved in fibrinogen binding and loops a3- a4 and bD-a5 are reported to be more important for recognition of fibrinogen. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service