Skip to Content
Merck
All Photos(11)

Key Documents

ZRB1266

Sigma-Aldrich

Anti-Na+-K+ ATPase α1 Antibody, clone 1E13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$239.00
4 X 25 μL
$492.00

$239.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$239.00
4 X 25 μL
$492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$239.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E13, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~112.90 kDa
observed mol wt ~100 kDa

species reactivity

human

species reactivity (predicted by homology)

bovine, rat, monkey, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E13 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Na(+)/K(+) ATPase a1 subunit. It targets an epitope within 16 amino acids from the third cytoplasmic domain from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the third cytoplasmic domain from the N-terminal half of human Na(+)/K(+) ATPase a1 subunit.

Application

Quality Control Testing

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Na+-K+ ATPase a1 in A431 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Na+-K+ ATPase 1 in MCF-7, A549. and Human colon lysates.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Na+-K+ ATPase 1 in A549 cell line.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Na+-K+ ATPase 1 in human colon tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Na+-K+ ATPase a1, clone 1E13 ZooMAb®, Cat. No. ZRB1266, is a rabbit recombinant monoclonal antibody that specifically targets of Na+-K+ ATPase a1 and is tested for use in Immunocytochemistry, Immunohistochemistry, and Western Blotting.

Target description

Sodium/potassium-transporting ATPase subunit alpha-1 (UniProt: P05023; also known as EC:7.2.2.13, Na (+)/K (+) ATPase alpha-1 subunit, Sodium pump subunit alpha-1) is encoded by the ATP1A1 gene (Gene ID: 476) in human. Na (+)/K (+) ATPase alpha-1 is a multi-pass membrane protein with five extracellular domains; ten helical domains, and six cytoplasmic domains. It is synthesized with a short propeptide (aa 1-5), which is removed in the mature form. It serves as the catalytic component of the active enzyme that catalyzes the hydrolysis of ATP coupled with the exchange of sodium and potassium ions across the plasma membrane. This action creates the electrochemical gradient of sodium and potassium ions that provides energy for active transport of various nutrients. This gradient is also essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. Phosphorylation of Na (+)/K (+) ATPase alpha-1 on tyrosine 10 modulates its pumping activity and phosphorylation on serine 943 by protein kinase A modulates its response protein kinase C. Following increase in intracellular sodium, it undergoes dephosphorylation by protein phosphatase 2A (PP2A) that leads to increase in its catalytic activity. Mutations in ATP1A1 gene have been linked to Charcot-Marie-Tooth disease, a disorder of the peripheral nervous system, characterized by progressive weakness and atrophy. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service