Skip to Content
Merck
All Photos(15)

Key Documents

ZRB1207

Sigma-Aldrich

Anti-CD19 Antibody, clone 3E7 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3E7, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 61.13 kDa
observed mol wt ~80 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminus

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3E7 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects CD19. It targets an epitope within 23 amino acids from the N-terminal, extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the N-terminal region of human CD19.

Application

Quality Control Testing

Evaluated by Western Blotting in Daudi cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CD19 in Daudi cell lysate.

Tested applications

Flow Cytometry Analysis: 1 μg from a representative lot detected CD19 in one million Human peripheral blood mononuclear cells (PBMC).

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CD19 in Raji cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CD19 in human tonsil and human appendix tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-CD19, clone 3E7 ZooMAb®, Cat. No. ZRB1207,is a recombinant Rabbit monoclonal antibody that specifically targets CD19 and is tested for use in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

B-lymphocyte antigen CD19 (UniProt: P15391; also known as B-lymphocyte surface antigen B4, Differentiation antigen CD19, T-cell surface antigen Leu-12, CD19) is encoded by the CD19 gene (Gene ID: 930) in human. CD19 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-19), which are subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 20-291), a transmembrane domain (aa 292-313), and a cytoplasmic domain (aa 314-556). It also contains two Ig-like C2-type domains (aa 20-113 and 176-277). CD19 expression has been reported in B cells in blood and lymph nodes. It is part of a complex that is composed of CD19, CD21, CD81, and CD225 in the membrane of mature B-cells. It interacts directly with CD81 via its second extracellular domain early during biosynthesis in the ER/pre-Golgi compartments and is essential for trafficking and compartmentalization of CD19 receptor on the cell surface of activated B cells. It functions as a coreceptor for the B-cell antigen receptor complex (BCR) on B-lymphocytes and is reported to decrease the threshold for activation of downstream signaling pathways and for triggering B-cell responses to antigens. Following B cell activation, CD19 is phosphorylated on tyrosine residues in a sequential order by Lyn. Phosphorylation of Tyr-531 is rapid, followed by phosphorylation at Tyr-409. Phosphorylation of Tyr-500 appears to be slow and transient. CD19 is shown to be essential for normal B cell differentiation and proliferation in response to antigen challenges. It also activates signaling pathways that lead to the activation of phosphatidylinositol 3-kinase and the mobilization of intracellular Ca2+ stores. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Sato, A., et al. (1997). J. Immunol. 159(7); 3278-3287; Roifman, CM., and Ke, S. (1992). Biochem. Biophys. Res. Commun. 194(1); 222-225; Carter, RH., and Fearon, DT (1992). Science. 256(5053); 105-107).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service