ZMS1209
Anti-PNPLA3 Antibody, clone 11C5 ZooMAb® Mouse Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
1-acylglycerol-3-phosphate O-acyltransferase PNPLA3, ADPN, Acylglycerol transacylase, Adiponutrin, Calcium-independent phospholipase A2-epsilon, EC:2.3.1.51, Lysophosphatidic acid acyltransferase, Patatin-like phospholipase domain-containing protein 3, iPLA2-epsilon
About This Item
Recommended Products
biological source
mouse
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
recombinant monoclonal
description
clone 11C5
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 52.86 kDa
observed mol wt ~52 kDa
purified by
using protein G
species reactivity
human
packaging
antibody small pack of 25
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG2aκ
epitope sequence
Unknown
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... PNPLA3(80339)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in HeLa cell lysate.
Western Blotting Analysis: A 1:10,000 dilution of this antibody detected PNPLA3 in HeLa cell lysate.
Tested Applications
Western Blotting Analysis: A 1:10,000 dilution of this antibody detected GST-tagged, full-length, recombinant Human PNPLA3.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected PNPLA3 in Human kidney tissue sections.
Enzyme Immunoassay (ELISA) Analysis: Serial dilutions from a representative lot detected GST-tagged, full-length, recombinant Human PNPLA3.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PNPLA3 in HepG2 cells.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service