Skip to Content
Merck
All Photos(9)

Documents

ZMS1165

Sigma-Aldrich

Anti-CHRM4 Antibody, clone 18C7.2 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-ZFP36L2 Antibody, clone 1E10 ZooMAb® Rabbit Monoclonal, Muscarinic M4 receptor, Muscarinic acetylcholine receptor M4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

18C7.2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 53.05 kDa
observed mol wt ~56 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Cytoplasmic domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... CHRM4(1132)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 18C7.2 is a ZooMAb® mouse recombinant monoclonal antibody that specifically detects Muscarinic acetylcholine receptor M4 (CHRM4). It targets an epitope within the third intracellular loop.

Immunogen

GST-tagged recombinant fragment corresponding to the third intracellular loop of human muscarinic acetylcholine receptor M4 (CHRM4).

Application

Quality Control Testing

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Muscarinic acetylcholine receptor M4 (CHRM4) in SHSY5Y cell lysate.

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound full-length recombinant Human CHRM4 protein with a KD of 5.1 x 10-6 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CHRM4 in SH-SY5Y cells.

Enzyme Immunoassay (ELISA) Analysis: A serial of dilutions from a representative lot detected full-length recombinant Human CHRM4.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Muscarinic acetylcholine receptor M4 (UniProt: also known as Muscarinic M4 receptor) is encoded by the CHRM4 gene (Gene ID: 1132) in human. Acetylcholine receptors are classified as muscarinic or nicotinic. Muscarinic receptors are found in neuroeffector junctions and are coupled to G-proteins. On the other hand, nicotinic receptors are found in the autonomic ganglia and neuromuscular junctions. Muscarinic receptors are G-protein-coupled receptors that are involved in the transduction of cholinergic signals in the central nervous system, autonomic ganglia, smooth muscles, and other parasympathetic end organs. Five different subtypes of muscarinic receptors have been described (M1-M5). The M1, M3, and M5 receptors are excitatory receptors, whereas M2 and M4 receptors inhibit neuronal firing by decreasing the intracellular concentration of cAMP and activating G-protein coupled inward rectifying potassium channels. M1, M3, and M5 receptors are coupled to Gq/11 protein, while M2 and M4 pre

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service