Skip to Content
Merck
All Photos(14)

Key Documents

ZMS1049

Sigma-Aldrich

Anti-Clusterin α chain Antibody, clone 41D ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$281.00
4 X 25 μL
$579.00

$281.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$281.00
4 X 25 μL
$579.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$281.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

41D, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 52.50 kDa
observed mol wt ~60 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 41D is a ZooMAb® mouse recombinant monoclonal antibody that specifically detects human Clusterin a.

Immunogen

Full length, purified, native Clusterin from human serum.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Clusterin a in HeLa cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Clusterin a chain in MCF-7 and PANC-1 cell lysates.

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected Clusterin a in human tonsil tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Clusterin a in human adrenal gland and human spleen tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Clusterin chain, clone 41D ZooMAb®, Cat. No. ZMS1049, is a recombinant Mouse monoclonal antibody that detects Clusterin and is tested for use in and Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Clusterin (UniProt: P10909; also known as Aging-associated gene 4 protein, Apolipoprotein J, Apo-J, Complement cytolysis inhibitor, CLI, Complement-associated protein SP-40,40, Ku70-binding protein 1, NA1/NA2, Sulfated glycoprotein 2, SGP-2, Testosterone-repressed prostate message 2, TRPM-2) is encoded by the CLU (also known as APOJ, CLI, KUB1, AAG4) gene (Gene ID: 1191) in human. Clusterin is a ubiquitously expressed protein that functions as extracellular chaperone that prevents aggregation of non-native proteins. It also prevents stress-induced aggregation of plasma proteins. Clusterin is synthesized with a signal peptide (aa 1-22), which is subsequently cleaved off to generate the mature form that can be proteolytically cleaved into a b-chain (aa 23-227) and an a-chain (aa 228-449). However, proteolytic cleavage is not necessary for its chaperone activity. Six different isoforms of Clusterin have been described that are produced by alternative splicing. Isoform 1 is shown to retro-translocate from the secretory compartments to the cytosol upon cellular stress, but isoforms 4 and 6 retain their cytoplasmic localization in stressed and unstressed cell. It maintains partially unfolded proteins in a state appropriate for subsequent refolding by other chaperones, such as HSPA8/HSC70, but does not refold proteins by itself. Clusterin is reported to bind to Amyloid b-peptides and alters their aggregation and promote their clearance, suggesting its neuroprotective role. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Foster, EM., et al. (2019). Front. Neurosci. 13; 64; Rohne, P., et al. (2014). Cell Physiol. Biochem. 34(5); 1626-1639; Amy R Wyatt, AR., and Wilson, MR. (2010). J. Biol. Chem. 285(6); 3532-3539; Poon, S., et al. (2000). Biochemistry. 39(51); 15953-15960).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service