Skip to Content
Merck
All Photos(1)

Key Documents

Z768744

Eppendorf Tube®

Biopur®, tube capacity 5 mL

Synonym(s):

centrifugation, centrifuge tubes, microtubes, sampling tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121702
NACRES:
NB.11

material

transparent polypropylene

feature

Biopur®
conical shaped bottom with hinged lid

packaging

pkg of 50 ea (individually wrapped)

manufacturer/tradename

Eppendorf® 0030 119.479 (European)
Eppendorf® 0030119479 (US)

parameter

25,000 × g (g-force rating)

tube capacity

5 mL

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf Tubes are the perfect option for working with medium-sized sample volumes!

  • Maximum safety and stability for centrifugation up to 25,000 × g
  • Conical bottoms
  • Easier processing of samples up to 5.0 mL
  • Higher yields in DNA isolation, prep of mastermixes and buffers, and safe cell and tissue lysis
  • Transparent polypropylene, free of plasticizers, biocides or mold release agents
  • Hinged lid for minimized sample evaporation during storage and incubation in a wide range of temperatures from -86°C to 80°C

Legal Information

Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Tube is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service