Z757969
Sartorius Picus electronic pipette
12 channel, volume 5-120 μL, pack of 1 ea
Synonym(s):
electronic pipetting, pipettors
Select a Size
$3,040.00
Select a Size
About This Item
$3,040.00
Recommended Products
AC/DC input
100 V / 240 V AC, European 2-pin plug (flat 2-pin)
100 V / 240 V AC, UK plug (3 pin)
100 V / 240 V AC, US 2-pin plug
feature
12-channel
CE compliant
electronic
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 735441
volume
5-120 μL
battery power
rechargeable
suitability
suitable for (Product is used in uHPLC, HPLC and trace analysis applications.)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Electronic tip ejection, one hand-operated adjustment wheel, and conveniently located start button minimizes the force needed in pipetting. Repetitive and time consuming pipetting can be done accurately and fast with serial dispensing modes and a new Tracker - great help for example in microplate work.
- Tracker in conjunction with pipetting, reverse pipetting and multi dispensing modes to help microplate work
- Adjustment possibility for 1-point, 2-points and 3-points
- Easy maintenance of lower parts
- Safe-Cone filters in all models > 10 μl to prevent contaminations
- Autoclavable lower parts in all models (excl. mcp 1200 μL)
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service