Z744270
Wilmad® Suprasil® Electrolytic Assembley
for use with Joel, pack of 1
Synonym(s):
EPR kinetic study assembly, electrolytic cell assembly
About This Item
Recommended Products
packaging
pack of 1
manufacturer/tradename
Wilmad WG-810-J-Q
compatibility
for use with Joel
Looking for similar products? Visit Product Comparison Guide
General description
JEOL spectrometers is 10mm outside (WG-810-J-Q), for other spectrometers, the cell width is 10.5mm outside (WG-810-Q). The Gas Bubbler Assembly is a three-piece Pyrex® chamber which holds part of your sample it has three Standard Taper 10/30 outer joints at the top for the Gas Bubbler and two Calomel Electrodes. The lower Standard Taper 10/30 inner joint fits into the top of the Flat Electrolytic Cell. Nitrogen should be bubbled through the solution before and during the experiment to remove paramagnetic oxygen and mix the sample during electrolysis. The Platinum Electrodes are prepared using 0.016? (0.4mm) diameter wire. The upper electrode, WG-810-C, uses 8? (203mm) of platinum and acts as the working electrode in oxidation reactions. The lower electrode, WG-810-D, has only 2? (50mm) of platinum and acts as the auxiliary electrode in oxidation reactions. If you add a few drops of mercury to the cell, the lower electrode can be used as the working cathode in reduction reactions. Each Platinum Electrode incorporates a Pt-to-W weld. The tungsten (W) external wire lets you quickly clip your potentiostat leads to the electrode.Electrolysis Limits in Water (vs. Saturated Calomel Electrode)
Application
Features and Benefits
- Complete cell includes two Calomel Reference Electrodes, a pair of Platinum Electrodes, and a gas bubbler
- Electrolytic Cell Assembly comes as a complete unit with CFQ or Suprasil® Flat Cell
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service