Z741974
Cameo™ Filtration Membrane
Nylon membrane, pore size 5.0 μm, diam. 47 mm, pkg of 100 ea
Select a Size
$287.00
Estimated to ship onJune 01, 2025
Select a Size
About This Item
$287.00
Estimated to ship onJune 01, 2025
Recommended Products
material
Nylon membrane
sterility
non-sterile
packaging
pkg of 100 ea
manufacturer/tradename
GVS Life Sciences 1213812
diam.
47 mm
pore size
5.0 μm
General description
Applications: Analysis of Biological and Organic Fluids, Analytical Sample Prep, HPLC, Composite assays, Content uniformity, Dissolution testing, Environmental samples, Food Analysis
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)