Key Documents
Z741757
4-liter Culture media bottle
Select a Size
1 EA
$393.00
$393.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$393.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
$393.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polycarbonate
feature
Comes with cap, tubing and fixtures.
manufacturer/tradename
IBI ACCFL0008
Related Categories
General description
The 4L culture media bottle is set up to provide source culture media for an IBI Flow Cell system. The bottle is made of polycarbonate and can be autoclaved. It comes complete with cap, containing fixtures and tubing to connect to the 3-channel or Convertible flow cell systems. - See more at: http://www.ibisci.com/product/accfl0008#sthash.hO4T0B0H.dpuf
Choose from one of the most recent versions:
Certificates of Analysis (COA)