Key Documents
Z741658
Pipetting Reagent Vessel
Select a Size
16 EA
$106.00
$106.00
Estimated to ship onMay 25, 2025
Select a Size
Change View
16 EA
$106.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
$106.00
Estimated to ship onMay 25, 2025
material
polypropylene
packaging
pack of 16
manufacturer/tradename
Sartorius 783500
capacity
120 mL
General description
Made of polypropylene, the autoclavable and durable reagent vessel is chemically resistant to all common reagents. It is especially useful with all multichannel pipettes.
Choose from one of the most recent versions:
Certificates of Analysis (COA)