Z741649
Sartorius pipette tips
volume range 100-5000 μL, Standard, rack, sterile
Select a Size
$60.50
Select a Size
About This Item
RNase free
barrier: no
endotoxin free
$60.50
Recommended Products
material
colorless
sterility
sterile
feature
DNase free
RNase free
barrier: no
endotoxin free
packaging
pack of 50 ea
manufacturer/tradename
Sartorius 780305
maximum volume
5 mL
volume range
100-5000 μL
volume
100—5000 μL
Related Categories
General description
Optifit Tips are available in various packaging types for all your needs:
- Single Tray racks – purity certified tip racks
- Refill Packs - purity certified and ecological refill packages
- Refill Towers - space-saving and ecological refill packages
- Flexibulk boxes - space-saving, ecological and economical bulk boxes
- Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)