Skip to Content
Merck
All Photos(1)

Key Documents

Z740423

Tamper proof specimen containers

size 2600 mL

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
$428.00

$428.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025



Select a Size

Change View
25 EA
$428.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

$428.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


material

polypropylene cap
polypropylene container

packaging

pack of 25 ea

manufacturer/tradename

Simport C581-2600

size

2600 mL

Looking for similar products? Visit Product Comparison Guide

General description

These disposable containers are ideal for collection, transport and storage of fluids, powders, solids, pathology specimens and hazardous samples. Tight-fitting lids prevent leaks and odors. They incorporate a tamperproof lid which is opened by first removing tab on side of lid, then by lifting the cover. They resist temperatures up to 80 °C. The two larger sizes are supplied with a plastic handle for easy carrying.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What are the dimensions for the 2.6 L container?

    1 answer
    1. The product dimensions are as follows:
      Opening diameter: 200 mm
      Height: 124 mm

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service