Skip to Content
Merck
All Photos(1)

Key Documents

Z721417

SealPlate® ColorTab films

green

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$189.00

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
100 EA
$189.00

About This Item

UNSPSC Code:
41122410
NACRES:
NB.22

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

polyester film

sterility

non-sterile

packaging

pack of 100 ea

manufacturer/tradename

Excel Scientific SP-IDG-100

technique(s)

ELISA: suitable

L

146 mm

W × L

7.94 cm × 14.6 cm

W

79.4 mm

size

Up to 1536 wells

color

green

Looking for similar products? Visit Product Comparison Guide

General description

SealPlate ColorTab adhesive films consist of 50 μm thickness clear polyester. They minimize evaporation, prevent spillage and contamination between wells, and provide a secure seal for ELISA, EIA, and similar assays. These unique films include a color dot and white writing area on each end tab for plate ID labeling. End tabs with backing removed adhere to the ends of the plate and are perforated so they can be left in place to maintain plate identity even if the center portion of the film is removed. Alternatively, one inscribed color-coded end tab can be removed and pasted in a laboratory notebook while the other remains on the plate for identification. Dimensions 79.4 by 146.0 mm for 96 well plates. Films are supplied with five dot colors (green, lavender, orange, red, blue) in non-sterile single-color packs of 100 and in assorted packs of 50 containing five individual-color packs of 10.

Legal Information

ColorTab is a trademark of Excel Scientific, Inc.
SealPlate is a registered trademark of Excel Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service