Key Documents
Z719560
Sorenson low binding E tips
maximum volume 10 μL, case of 4800 ea (50 racks of 96 tips), sterile
Select a Size
960 EA
$310.00
4800 EA
$1,180.00
$310.00
Estimated to ship onMay 22, 2025
Select a Size
Change View
960 EA
$310.00
4800 EA
$1,180.00
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
10 μL
sterility:
sterile
feature:
barrier: no
packaging:
case of 4800 ea (50 racks of 96 tips)
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Sorenson 22210T
$310.00
Estimated to ship onMay 22, 2025
material
colorless
polypropylene
sterility
sterile
feature
barrier: no
packaging
case of 4800 ea (50 racks of 96 tips)
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Sorenson 22210T
maximum volume
10 μL
volume
10 μL
General description
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Extended length. Fits Eppendorf ultra-micro pipettors.
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)