Key Documents
Z719552
Sorenson low binding E tips
maximum volume 10 μL, non-sterile
Select a Size
960 EA
$96.60
4800 EA
$359.00
$96.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
960 EA
$96.60
4800 EA
$359.00
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
10 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
case of 4800 ea (50 racks of 96 tips)
pack of 96 ea (Racks)
manufacturer/tradename:
Sorenson 22220T
$96.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
colorless
polypropylene
sterility
non-sterile
feature
barrier: no
packaging
case of 4800 ea (50 racks of 96 tips)
pack of 96 ea (Racks)
manufacturer/tradename
Sorenson 22220T
maximum volume
10 μL
volume
10 μL
Related Categories
General description
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Extended length. Fits Eppendorf ultra-micro pipettors.
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)