Select a Size
$1,200.00
Estimated to ship onApril 13, 2025
Select a Size
About This Item
$1,200.00
Estimated to ship onApril 13, 2025
Recommended Products
feature
electronic: no
single-channel
packaging
pack of 1 ea
pkg of 1 ea
manufacturer/tradename
Nichiryo 00-NPL02-2
availability
not available in (Not available in France)
volume
0.1-2.0 μL
General description
Time and money spent on the calibration of pipettes is now greatly reduced!
- UV resistant
- Solvent resistant
- Fully autoclavable (121°C for 20 minutes)
- One-touch calibration
- Ceramic pistons
- Filtered nozzles
- Easy, convenient locking lever
- Ergonomic design
- Single channel, manual, continuously adjustable volume
- Advanced durability against organic solvent dispensing
- PTFE (Fluorine resin) in the air tight chamber of the instrument
- A Perfluoro rubber O-ring with low cubical expansion in the airtight parts
- Plunger made from a material resistant to organic solvents
- An alloy spring to avoid metal corrosion by organic solvents
- Quick and easy to use
Choose from one of the most recent versions:
Certificates of Analysis (COA)