Key Documents
Z710857
Silicone membrane liner for Tunair shake flask
For 300 mL flask
Select a Size
5 EA
$450.00
$450.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
5 EA
$450.00
About This Item
UNSPSC Code:
41122104
NACRES:
NB.22
$450.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
silicon
sterility
non-sterile
packaging
case of 5
manufacturer/tradename
IBI SS-3016
General description
Silicone membrane liner for Tunair shake flask consists of five 0.22 m silicone membrane cap linings. The filter lining in the cap assembly is replaced by tweaking the flanges of the inner-closure shell until they separate loose. Then it is pulled apart to remove the used lining. The lining can then be replaced by sandwiching it between two parts of the cap and the cap is snapped back together. While reassembling, ensure the flanges from the inner piece fits into the mated grooves in the outer piece. It is required for the cap assembly to stay intact while using.
Features and Benefits
  • Long-lasting unless punctured
  • Rinse with water only if visibly soiled.
  • Recommends autoclaving cap along with the filter.
Legal Information
Tunair is a trademark of IBI Scientific
Choose from one of the most recent versions:
Certificates of Analysis (COA)