Skip to Content
Merck
All Photos(1)

Key Documents

Z709263

ART® self-sealing barrier pipette tips

ART 300, volume range 1-300 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
volume:
1—300 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Molecular BioProducts 2070
Pricing and availability is not currently available.

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier

packaging

pack of 960 ea (10 racks of 96 tips)

manufacturer/tradename

Molecular BioProducts 2070

technique(s)

PCR: suitable

maximum volume

300 μL

volume range

1-300 μL

volume

1—300 μL

compatibility

for use with

  • sku=2070
  • Biohit Electronic 10-250 μl
  • Biohit Electronic Multi-Channel 25-250 μl
  • Biohit Variable 50-250 μl
  • Eppendorf Response 4850 20-500 μl
  • Eppendorf Research Pro 4860 20-300 μl
  • Eppendorf Titerman 4908 30-300 μl
  • Finnpipette Multi-Channel 50-300 μl
  • Hamilton SoftGrip Precision 300 μl
  • Rainin EDP 2 20-250 μl
  • Rainin EDP Plus 20-250 μl
  • Socorex/Wheaton Calibra/822 20-200 μl
  • Transferpette Multi-Channel 20-200ul/30-300 μl

Looking for similar products? Visit Product Comparison Guide

General description

A secure fit and extra space for repeat pipetting

Application

ART® self-sealing barrier pipette tips have been used for the quantitative profiling of endocannabinoids in lipoproteins by LC–MS (liquid chromatography–mass spectrometry)/MS.[1] They have also been used for RNA isolation and reverse transcription.[2]

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro culture and analysis of adult hippocampal neural progenitors.
Peltier J, et al.
Methods in Molecular Biology, 621, 65-87 (2010)
Quantitative profiling of endocannabinoids in lipoproteins by LC-MS/MS.
Bilgin M, et al.
Analytical and Bioanalytical Chemistry, 407, 5125-5131 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service