Key Documents
Z709034
ART® non-filtered tip
HLT 200 Micropoint, size 0.5-200 μL, Pure packed, sterile
Synonym(s):
pipet tips, pipette tips
Select a Size
1 PKG
$206.00
5 PKG
$763.00
$206.00
Estimated to ship onMay 28, 2025
Select a Size
Change View
1 PKG
$206.00
5 PKG
$763.00
About This Item
UNSPSC Code:
41121600
eCl@ss:
32079201
NACRES:
NB.01
volume:
0.5—200 μL
sterility:
sterile
feature:
DNase free
RNase free
Pure packed
packaging:
pack of 960 ea
manufacturer/tradename:
Molecular BioProducts 3551
$206.00
Estimated to ship onMay 28, 2025
sterility
sterile
feature
DNase free
RNase free
Pure packed
packaging
pack of 960 ea
manufacturer/tradename
Molecular BioProducts 3551
maximum volume
200 μL
size
0.5-200 μL
volume
0.5—200 μL
General description
Micropoint design and 10 μl reference mark for accurate sample handling.
Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)