Key Documents
Z708410
Refrigerator rack
Holds 50 x 15 or 50 mL tubes
About This Item
UNSPSC Code:
41122806
NACRES:
NB.22
material
polystyrene
feature
Holds 50 x 15 or 50 mL tubes
manufacturer/tradename
Heathrow Scientific HS28230B
technique(s)
cryopreservation: suitable
size
276x164x134 mm
General description
Quick chill sample tubes. Two-piece rack has a lower section tray which can hold water or crushed ice. Top insert tray secures tubes. Molded polystyrene racks are designed to stack on top of each other to save benchtop space.
Choose from one of the most recent versions:
Certificates of Analysis (COA)