Skip to Content
Merck
All Photos(1)

Key Documents

Z708356

Disposable forceps

dark blue, L 110 mm, straight tip

Synonym(s):

single use forceps, single use tweezers

Sign Into View Organizational & Contract Pricing

Select a Size

6 EA
$76.10

$76.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
6 EA
$76.10

About This Item

UNSPSC Code:
41122405
eCl@ss:
32035504
NACRES:
NB.35

$76.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

manufacturer/tradename

Heathrow Scientific HS15998C

L

110 mm

color

dark blue

Looking for similar products? Visit Product Comparison Guide

General description

Disposable forceps feature corrugated handle and serrated tips. The yellow forceps are made of polystyrene, both blue forceps are acrylonitrile butadiene styrene.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service