Key Documents
Z694479
Nanofiber chamber slides
8 chamber slide, random oriented nanofibers
Synonym(s):
3D, 3D Cell Culture, 3d plates, tissue culture plates
Select a Size
1 EA
$99.60
$99.60
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onMay 31, 2025
Select a Size
Change View
1 EA
$99.60
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
sterility:
non-sterile
$99.60
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onMay 31, 2025
sterility
non-sterile
manufacturer/tradename
Nanofiber Solutions 801
suitability
suitable for (Tissue Engineering, Cancer Research)
General description
Nanofiber cell culture dishes, multiwell plates, chamber slides and well plate inserts combine the best of 3D and 2D cell culture. The nanofibers are optically transparent to allow for live-cell imaging and real time quantification of cell mobility. The nanofibers mimic the 3D topography in vivo, and can be easily coated with ECM proteins for Cancer Research. The Cells can be simply removed for protein or gene analysis.
The nanofibers have higher expansion rates, better differentiation control, and has a synthetic surface with no animal contamination, ideal for stem cells research and immunohistochemistry staining. Nanofiber scaffolds can be easily shaped or sized, made from nearly any synthetic or natural polymer for customized tissue engineering. They can then be implanted in vivo or used in vitro. The standard plate dimensions available will work with current automated equipment for high throughput drug discovery needs.
The cell culture dishes, plates, chamber slides and inserts are available with either randomly oriented fibers or aligned fibers.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. What is the material used in this device?
    1 answer
    1. The fibers are made of polycaprolactone (PCL), a biomimetic polymer that resembles soft tissue, such as cartilage.
      Helpful?