Skip to Content
Merck
All Photos(1)

Documents

Z660051

Bandelin TICKOPUR R 27 cleaning concentrate

special acid cleaner for ultrasonic bath

Synonym(s):

low foaming detergent, ultrasonic bath detergent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.35

packaging

pkg of 1 L bottle
pkg of 2 L bottle
pkg of 5 L cannister

manufacturer/tradename

Bandelin 815

availability

available only in EU

General description

Special acid cleaner for ultrasonic bath, concentrate - based on phosphoric acid, anti-corrosive, acid, pH 1.9 (1%), dosage 5%, 1 to 10 min.

Contamination:

Heavy mineral residues like chalk, silicate, phosphate, rust, cement, temper colors, metal oxides, grease and oil films etc.

Objects to be cleaned:

Glass, ceramics, plastics, rubber, steel, stainless steel, precious metals. Not for light and non-ferrous metals, tin and zinc.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service