Z659746
Bandelin SONOREX™ SUPER without heating Ultrasonic baths
Bandelin RK 510, capacity 9.7 L
Synonym(s):
ultrasonic bath sonicator, ultrasonic cleaning bath
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
AC/DC input
230 V
feature
CE compliant
HF-peak output max. W/per = 2 × 320
rain ball cock = ½"
packaging
pack of 1 ea
manufacturer/tradename
Bandelin RK 510
availability
not available in USA and Canada
L × W × D
300 mm × 240 mm × 150 mm
capacity
9.7 L
Looking for similar products? Visit Product Comparison Guide
General description
PZT-large area transducers produce the high-powered ultrasonic intensity throughout the entire oscillating tank. Concentrated ultrasonic energy without irritating surface waves. Constant power and automatic frequency control always ensure optimum distribution of ultrasonic energy and reproducible results, regardless of the sizes of the pieces to be cleaned. Especially intense due to the advantageous frequency of 35 kHz.
- PZT-large area oscillating systems
- HF-Frequency 35 kHz
- Timer 1-15 minutes and continuous operation
- Oscillating tank of stainless steel
- Housing of stainless steel - drip proof
- Drain cock included for easy emptying (from RK 106 upward)
- SONOREX SUPER units are RFI-proof and CE marked
Legal Information
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service