Key Documents
Z659495
Bandelin SONOREX Ultrasonic bath accessories
positioning lid fits Z659460
Select a Size
1 EA
$44.60
$44.60
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Select a Size
Change View
1 EA
$44.60
About This Item
UNSPSC Code:
41103706
NACRES:
NB.74
$44.60
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
material
stainless steel
feature
for 2 beakers
packaging
pack of 1 ea
manufacturer/tradename
Bandelin DE 08
availability
not available in USA and Canada
General description
When using chemically aggressive liquids (e.g. TICKOPUR J 80 U), removing chemically aggressive or abrasive contamination or sonicating sample liquids, insert cups are used (indirect sonication). Because nothing may be placed on the bottom of the ultrasonic bath use a suitable perforated lids/insert strips to hold the insert cups. Several insert cups can be sonicated with different liquids at the same time.
Features and Benefits
  • For holding insert cups
  • Sound-absorbing
  • Shortening the heating phase
Legal Information
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)