Skip to Content
Merck
All Photos(1)

Key Documents

Z640271

Eppendorf® epT.I.P.S. box

volume range 100-5000 μL

Synonym(s):

pipetting tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01
volume:
100—5000 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 24 (includes 1 reusable box)
manufacturer/tradename:
Eppendorf® 0030 073.169
Eppendorf® 022491385
Pricing and availability is not currently available.

material

violet rack

sterility

non-sterile

feature

barrier: no

packaging

pack of 24 (includes 1 reusable box)

manufacturer/tradename

Eppendorf® 0030 073.169
Eppendorf® 022491385

L

120 mm

maximum volume

5 mL

volume range

100-5000 μL

volume

100—5000 μL

Looking for similar products? Visit Product Comparison Guide

General description

Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays, with plastic lids protecting the trays from contact during transfer to the working box.
Autoclavable working box, optimized for multichannel pipettes, with removable lid and larger footprint to avoid tipping. Stackable. Supplied full of tips.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

A step-by-step protocol for sample preparation, calibration, digestion, and inductively coupled plasma-mass spectrometric (ICP-MS) analysis of heavy metals in six different hemp-containing beverages.

Analysis of heavy metals and metals in cannabis and hemp containing personal care products and cosmetics include sample preparation by grinding, preparation of calibration solutions for a standard addition approach, a microwave digestion protocol and subsequent ICP-MS analysis.

Protocols

Complete workflow for the comprehensive analysis of terpenes in cannabis

A complete SEC-UV workflow for quantifying monoclonal antibody fragments, monomers, and aggregates, including procedure for a system suitability test and heat stressing of antibodies.

HPLC separation of 17 important cannabinoids including CBD, delta 9 THC and THCA. Read the application note.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service