Key Documents
Z640204
Eppendorf® epT.I.P.S. box
volume range 0.1-20 μL
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
0.1—20 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 96 (includes 1 reusable box)
manufacturer/tradename:
Eppendorf® 0030 073.029
Eppendorf® 022491318
Pricing and availability is not currently available.
material
medium gray rack
sterility
non-sterile
feature
barrier: no
packaging
pack of 96 (includes 1 reusable box)
manufacturer/tradename
Eppendorf® 0030 073.029
Eppendorf® 022491318
L
40 mm
maximum volume
20 μL
volume range
0.1-20 μL
volume
0.1—20 μL
General description
Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays, with plastic lids protecting the trays from contact during transfer to the working box.
Autoclavable working box, optimized for multichannel pipettes, with removable lid and larger footprint to avoid tipping. Stackable. Supplied full of tips.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Chromolith® RP-18e columns optimize Oligo Standard 6 separation with varied flow rates and ion-pairing reagent evaluation.
A step-by-step protocol for sample preparation, calibration, digestion, and inductively coupled plasma-mass spectrometric (ICP-MS) analysis of heavy metals in six different hemp-containing beverages.
Analysis of heavy metals and metals in cannabis and hemp containing personal care products and cosmetics include sample preparation by grinding, preparation of calibration solutions for a standard addition approach, a microwave digestion protocol and subsequent ICP-MS analysis.
LC-UV-MS workflow details teriparatide peptide mapping, including enzymatic digestion, separation conditions, and QTOF mass spectrometer identification.
See All
Protocols
A complete SEC-UV workflow for quantifying monoclonal antibody fragments, monomers, and aggregates, including procedure for a system suitability test and heat stressing of antibodies.