Skip to Content
Merck
All Photos(1)

Key Documents

Z637319

Pursept®-A disinfectant

1 L bottles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.35

manufacturer/tradename

BRAND 44830

Looking for similar products? Visit Product Comparison Guide

General description

Ingredients: 100 g Pursept® AF contain: 12.5 g didecyl-dimethyl-ammoniumchloride, 10.4 g alkylpropylenediaminebis-guanidiniumdiacetate, 1.5 g bis(aminopropyl)-laurylamine, 5 - 15% NTA, < 5% non-ionic surfactants, auxiliary agent.

Features and Benefits

  • Suitable for disinfecting sensitive communication equipment
  • Dermatologically tested
  • Ready for use

Other Notes

BR44830 (India)

For atomizer for the 1L bottles; please order Z637327 separately.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Pursept is a registered trademark of Merz GmbH & Co. KG

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.4 °F

Flash Point(C)

23 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service